Search
Search
| Gene Symbol: Thap7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105863.2 | NP_001099333.2 | 3 - 4 | 271 | 70 | |
| XM_008768864.1 | XP_008767086.1 | 2 - 3 | 254 | 70 | ||
| GenBank mRNA | AW916308.1 | - | 3 - 4 | 271 | 70 | |
| BC168998.1 | - | 2 - 3 | 193 | 7387094 | 70 | |
Set Membership: |
|