Search
Search

Atp13a5| Gene Symbol: Atp13a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191657.1 | NP_001178586.1 | 29 - 30 | 3507 | 71 | |
| XM_008768800.1 | XP_008767022.1 | 29 - 30 | 3532 | 71 | ||
Set Membership: |
|