Search
Search
| Gene Symbol: Dgkd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006245504.2 | XP_006245566.2 | 26 - 27 | 3215 | 58 | |
| XM_006245505.2 | XP_006245567.2 | 26 - 27 | 3215 | 58 | ||
| GenBank mRNA | FQ218774.1 | - | 6 - 7 | 714 | 58 | |
Set Membership: |
|