Search
Search

Col11a2
LOC100911886| Gene Symbol: Col11a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212528.1 | NP_997693.1 | 4 - 5 | 813 | 65 | |
| XM_006255928.2 | XP_006255990.1 | 4 - 5 | 1661 | 65 | ||
| XM_006255929.2 | XP_006255991.1 | 4 - 5 | 1661 | 65 | ||
| XM_008772720.1 | XP_008770942.1 | 4 - 5 | 1662 | 65 | ||
| XM_008772721.1 | XP_008770943.1 | 4 - 5 | 1767 | 65 | ||
| Gene Symbol: LOC100911886 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772788.1 | XP_008771010.1 | 4 - 5 | 541 | 65 | |
Set Membership: |
|