Search
Search
| Gene Symbol: Dmtn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108385.1 | NP_001101855.1 | 10 - 11 | 1182 | 82 | |
| XM_006252279.2 | XP_006252341.1 | 9 - 10 | 1317 | 82 | ||
| XM_006252280.2 | XP_006252342.1 | 9 - 10 | 1315 | 82 | ||
| XM_006252281.1 | XP_006252343.1 | 7 - 8 | 670 | 82 | ||
| XM_008770830.1 | XP_008769052.1 | 10 - 11 | 1180 | 82 | ||
| XM_008770831.1 | XP_008769053.1 | 10 - 11 | 1837 | 82 | ||
| GenBank mRNA | BC166453.1 | - | 9 - 10 | 1191 | 8374835 | 82 |
Set Membership: |
|