Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130779.2 | NP_570135.2 | 16 - 17 | 3019 | 121 | |
XM_006239847.2 | XP_006239909.1 | 15 - 16 | 2755 | 121 | ||
XM_006239848.2 | XP_006239910.1 | 15 - 16 | 2758 | 121 | ||
XM_006239849.2 | XP_006239911.1 | 15 - 16 | 2752 | 121 | ||
XM_006239850.2 | XP_006239912.1 | 15 - 16 | 2758 | 121 | ||
XM_008764532.1 | XP_008762754.1 | 16 - 17 | 2875 | 121 | ||
GenBank mRNA | M55075.1 | - | 16 - 17 | 2944 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |