Search
Search

Olfml2b| Gene Symbol: Olfml2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107195.1 | NP_001100665.1 | 2 - 3 | 842 | 66 | |
| XM_006250219.2 | XP_006250281.1 | 2 - 3 | 857 | 66 | ||
Set Membership: |
|