Search
Search
| Gene Symbol: Trim23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100637.1 | NP_001094107.1 | 3 - 4 | 456 | 67 | |
| XM_008760683.1 | XP_008758905.1 | 4 - 5 | 537 | 67 | ||
| XM_008760684.1 | XP_008758906.1 | 4 - 5 | 537 | 67 | ||
| GenBank mRNA | FM052834.1 | - | 3 - 4 | 440 | 67 | |
| L04760.1 | - | 3 - 4 | 307 | 67 | ||
Set Membership: |
|