Search
Search

FAM187A| Gene Symbol: FAM187A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013861.1 | NP_001013883.1 | 1 | 1268 | 97 | |
| GenBank mRNA | BC079299.1 | - | 1 | 1268 | 7134519 | 97 |
Set Membership: |
|