Search
Search

LOC302999| Gene Symbol: LOC302999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750793.3 | XP_003750841.1 | 1 - 2 | 1113 | 76 | |
| XM_006246070.2 | XP_006246132.1 | 2 - 3 | 1801 | 76 | ||
Set Membership: |
|