Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01754645_m1
          See other ITGAL GE Assays ›
          Gene Symbol:
          Itgal
          Gene Name:
          integrin, alpha L
          Gene Aliases:
          LFA-1
          Chromosome Location:
          Chr.1: 198744051 - 198781745 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01754645_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Itgal
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033998.2 NP_001029170.1 30 - 31 3401 77
          XM_006230269.2 XP_006230331.1 30 - 31 3545 77
          GenBank mRNA BC101849.1 - 30 - 31 3401 7938065 77
          CV076006.1 - 2 - 3 175 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002629.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Itgal

          Panther Classification:

          Molecular Function -

          integrin

          Gene Ontology Categories:

          Function(s) Process(es)

          T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
          heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
          leukocyte cell-cell adhesion
          cell-matrix adhesion
          cell surface receptor signaling pathway
          integrin-mediated signaling pathway
          single organismal cell-cell adhesion
          regulation of cell-cell adhesion
          positive regulation of cell-cell adhesion
          positive regulation of T cell proliferation
          receptor clustering
          activated T cell proliferation
          positive regulation of calcium-mediated signaling
          ICAM-3 receptor activity
          protein complex binding
          protein heterodimerization activity
          cell adhesion molecule binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P452222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687970 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20440 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164870 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P452222 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P452222 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P452222 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P402655 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P452222 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79658 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42334 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10711026 - -
          Affymetrix Rat Gene 1.0 ST Array 10711043 - -
          Affymetrix Rat Gene 1.0 ST Array 10711046 - -
          Affymetrix Rat Gene 1.0 ST Array 10711040 - -
          Affymetrix Rat Gene 1.0 ST Array 10711018 - -
          Affymetrix Rat Gene 1.0 ST Array 10711050 - -
          Affymetrix Rat Gene 1.0 ST Array 10711023 - -
          Affymetrix Rat Gene 1.0 ST Array 10711044 - -
          Affymetrix Rat Gene 1.0 ST Array 10711031 - -
          Affymetrix Rat Gene 1.0 ST Array 10711038 - -
          Affymetrix Rat Gene 1.0 ST Array 10711032 - -
          Affymetrix Rat Gene 1.0 ST Array 10711039 - -
          Affymetrix Rat Gene 1.0 ST Array 10711019 - -
          Affymetrix Rat Gene 1.0 ST Array 10711034 - -
          Affymetrix Rat Gene 1.0 ST Array 10711041 - -
          Affymetrix Rat Gene 1.0 ST Array 10711048 - -
          Affymetrix Rat Gene 1.0 ST Array 10711030 - -
          Affymetrix Rat Gene 1.0 ST Array 10711036 - -
          Affymetrix Rat Gene 1.0 ST Array 10711033 - -
          Affymetrix Rat Gene 1.0 ST Array 10711049 - -
          Affymetrix Rat Gene 1.0 ST Array 10711029 - -
          Affymetrix Rat Gene 1.0 ST Array 10711024 - -
          Affymetrix Rat Gene 1.0 ST Array 10711022 - -
          Affymetrix Rat Gene 1.0 ST Array 10711020 - -
          Affymetrix Rat Gene 1.0 ST Array 10711047 - -
          Affymetrix Rat Gene 1.0 ST Array 10711042 - -
          Affymetrix Rat Gene 1.0 ST Array 10711028 - -
          Affymetrix Rat Gene 1.0 ST Array 10711027 - -
          Affymetrix Rat Gene 1.0 ST Array 10711025 - -
          Affymetrix Rat Gene 1.0 ST Array 10711037 - -
          Affymetrix Rat Gene 1.1 ST Array 10711017 - -
          Rat Genome 230 2.0 Array 1393347_PM_at - -
          Rat Genome 230 2.0 Array 1393347_at - -
          Affymetrix HT Rat Focus Array 1393347_at - -
          Affymetrix HT Rat Focus Array 1393347_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100020432.rn.1 TC0100002629.rn.1 RTA TC
          Rat Expression Set 230 1393347_at - -
          Rat Expression Set 230 1393347_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10711017 - -
          Rat Genome U34 Set rc_AA858880_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21056744 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349131 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline