Search
Search
| Gene Symbol: Hcfc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139507.1 | NP_001132979.1 | 1 - 2 | 1316 | 59 | |
| XM_006229593.2 | XP_006229655.1 | 1 - 2 | 1295 | 59 | ||
| XM_006229594.2 | XP_006229656.1 | 1 - 2 | 1294 | 59 | ||
| XM_006229596.2 | XP_006229658.1 | 1 - 2 | 1292 | 59 | ||
| XM_006229597.1 | XP_006229659.1 | 1 - 2 | 1311 | 59 | ||
Set Membership: |
|