Search
Search

LOC501406| Gene Symbol: LOC501406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006227820.2 | XP_006227882.1 | 2 - 3 | 186 | 116 | |
| XM_006227821.2 | XP_006227883.2 | 2 - 3 | 351 | 116 | ||
Set Membership: |
|