Search
Search
| Gene Symbol: Eddm3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178103.2 | NP_835204.1 | 1 | 248 | 101 | |
| XM_008770507.1 | XP_008768729.1 | 2 | 761 | 101 | ||
| GenBank mRNA | AJ515238.1 | - | 1 | 248 | 101 | |
Set Membership: |
|