Search
Search
| Gene Symbol: Klf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037354.1 | NP_001032431.1 | 2 - 3 | 348 | 86 | |
| XM_006239984.2 | XP_006240046.1 | 2 - 3 | 474 | 86 | ||
| GenBank mRNA | DQ232764.2 | - | 2 - 3 | 348 | 86 | |
Set Membership: |
|