Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCGAGAAGAATGATTTTAATAA[A/G]TGCCAGGTTTCCTGTAAACGAGTGA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
LRRC34 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LRRC34 - leucine rich repeat containing 34 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRIQ4 - leucine rich repeats and IQ motif containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080460.1 | Intron | NP_001073929.1 | ||||
XM_005247421.2 | Intron | XP_005247478.1 | ||||
XM_006713613.3 | Intron | XP_006713676.1 | ||||
XM_011512748.1 | Intron | XP_011511050.1 | ||||
XM_011512749.1 | Intron | XP_011511051.1 |