Search
Search

ZNF541TAAGCCCCCTGAGGGCAGGGATTTT[A/C]TCTCAATCACAAGTGTAAGCCATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF541 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| ZNF541 - zinc finger protein 541 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001277075.1 | Intron | NP_001264004.1 | ||||
| XM_005259311.4 | Intron | XP_005259368.1 | ||||
| XM_005259314.3 | Intron | XP_005259371.1 | ||||
| XM_011527367.2 | Intron | XP_011525669.1 | ||||
| XM_011527368.2 | Intron | XP_011525670.1 | ||||
| XM_011527369.2 | Intron | XP_011525671.1 | ||||
| XM_011527370.2 | Intron | XP_011525672.1 | ||||
| XM_011527371.2 | Intron | XP_011525673.1 | ||||
| XM_011527372.2 | Intron | XP_011525674.1 | ||||
| XM_011527373.2 | Intron | XP_011525675.1 | ||||
| XM_011527374.2 | Intron | XP_011525676.1 | ||||
| XM_011527375.1 | Intron | XP_011525677.1 | ||||
| XM_017027351.1 | Intron | XP_016882840.1 | ||||