Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATTGACATTTAGGGATGTGGCC[A/G]TAGAATTCTCTCTGAAGGAGTGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603899 | ||||||||||||||||||||
Literature Links: |
ZNF85 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF85 - zinc finger protein 85 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256171.1 | 205 | Intron | NP_001243100.1 | |||
NM_001256172.1 | 205 | Intron | NP_001243101.1 | |||
NM_001256173.1 | 205 | Intron | NP_001243102.1 | |||
NM_003429.4 | 205 | Intron | NP_003420.2 | |||
XM_011528263.2 | 205 | Missense Mutation | ATA,GTA | I,V 11 | XP_011526565.1 | |
XM_011528264.1 | 205 | Intron | XP_011526566.1 | |||
XM_017027234.1 | 205 | Intron | XP_016882723.1 |