Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGCACTAACCAGTACATTATGC[A/C]CATCTGTTCTTTGTTCCCAAAACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609413 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERCC6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERCC6 - ERCC excision repair 6, chromatin remodeling factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000124.3 | 879 | Intron | NP_000115.1 | |||
NM_001277058.1 | 879 | Missense Mutation | GGG,GTG | G,V 676 | NP_001263987.1 | |
NM_001277059.1 | 879 | Missense Mutation | GGG,GTG | G,V 676 | NP_001263988.1 |
PGBD3 - piggyBac transposable element derived 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_170753.3 | 879 | Missense Mutation | GGG,GTG | G,V 208 | NP_736609.2 |