Search
Search

CXCL16
MED11
ZMYND15AGCTGCCAGACTGTGGCCCGCAGTG[G/T]GAATGGTGGTTTCATTGGGACGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
CXCL16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| CXCL16 - C-X-C motif chemokine ligand 16 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001100812.1 | 1107 | Missense Mutation | CAC,CCC | H,P 192 | NP_001094282.1 | |
| NM_022059.3 | 1107 | Missense Mutation | CAC,CCC | H,P 192 | NP_071342.2 | |
| MED11 - mediator complex subunit 11 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| ZMYND15 - zinc finger MYND-type containing 15 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||