Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTAATTAATATGATCTATACTTAC[A/G]TCATCCTTGCAGATTTCAGTAATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610316 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PNPT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PNPT1 - polyribonucleotide nucleotidyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033109.4 | 2100 | Silent Mutation | GAC,GAT | D,D 671 | NP_149100.2 | |
XM_005264629.2 | 2100 | Intron | XP_005264686.1 | |||
XM_017005172.1 | 2100 | Intron | XP_016860661.1 |