Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAATGAACAAAACTTTGTGTCCA[A/C]ATCAGATATCAAGTCCATGTCTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610661 MIM: 610324 | ||||||||||||||||||||
Literature Links: |
NGLY1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NGLY1 - N-glycanase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OXSM - 3-oxoacyl-ACP synthase, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145391.1 | 433 | Missense Mutation | AAA,ACA | K,T 109 | NP_001138863.1 | |
NM_017897.2 | 433 | Missense Mutation | AAA,ACA | K,T 109 | NP_060367.1 | |
XM_006713216.3 | 433 | Missense Mutation | AAA,ACA | K,T 109 | XP_006713279.1 | |
XM_006713217.3 | 433 | Intron | XP_006713280.1 | |||
XM_017006713.1 | 433 | Missense Mutation | AAA,ACA | K,T 109 | XP_016862202.1 | |
XM_017006714.1 | 433 | Intron | XP_016862203.1 |