Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTAGCTACTGCTTTAGCAAGGAG[C/T]GTCTTCCCCGTGCCAGGTGGGCCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606696 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KATNA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KATNA1 - katanin catalytic subunit A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204076.1 | 1117 | Silent Mutation | ACC,ACT | T,T 180 | NP_001191005.1 | |
NM_007044.3 | 1117 | Silent Mutation | ACC,ACT | T,T 256 | NP_008975.1 | |
XM_005266804.2 | 1117 | Silent Mutation | ACC,ACT | T,T 256 | XP_005266861.1 | |
XM_005266805.3 | 1117 | Intron | XP_005266862.1 | |||
XM_017010207.1 | 1117 | Silent Mutation | ACC,ACT | T,T 256 | XP_016865696.1 | |
XM_017010208.1 | 1117 | Silent Mutation | ACC,ACT | T,T 128 | XP_016865697.1 | |
XM_017010209.1 | 1117 | Silent Mutation | ACC,ACT | T,T 128 | XP_016865698.1 | |
XM_017010210.1 | 1117 | Silent Mutation | ACC,ACT | T,T 256 | XP_016865699.1 |
RPS18P9 - ribosomal protein S18 pseudogene 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |