Search
Search

ELK1GTACCTTGTCATAGTAGTACCGCAA[A/G]GCCCGGCTGAGCTTGTCGTAATTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
ELK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| ELK1 - ELK1, ETS transcription factor | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001114123.2 | 511 | Silent Mutation | GCC,GCT | A,A 63 | NP_001107595.1 | |
| NM_001257168.1 | 511 | Silent Mutation | GCC,GCT | A,A 63 | NP_001244097.1 | |
| NM_005229.4 | 511 | Silent Mutation | GCC,GCT | A,A 63 | NP_005220.2 | |
| XM_017029339.1 | 511 | Silent Mutation | GCC,GCT | A,A 63 | XP_016884828.1 | |