Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCTGGAGAACGAGCTGAGTGGGA[A/G]CAGTAGTACTGGCAGGAGGCCCATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300688 MIM: 300775 | ||||||||||||||||||||
Literature Links: |
BCORL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BCORL1 - BCL6 corepressor-like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ELF4 - E74 like ETS transcription factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127197.1 | 2034 | Missense Mutation | GCT,GTT | A,V 438 | NP_001120669.1 | |
NM_001421.3 | 2034 | Missense Mutation | GCT,GTT | A,V 438 | NP_001412.1 | |
XM_005262389.3 | 2034 | Missense Mutation | GCT,GTT | A,V 438 | XP_005262446.1 | |
XM_011531307.2 | 2034 | Missense Mutation | GCT,GTT | A,V 370 | XP_011529609.1 | |
XM_011531308.2 | 2034 | Missense Mutation | GCT,GTT | A,V 361 | XP_011529610.1 |