Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMB8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| PSMB8 - proteasome subunit beta 8 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_004159.4 | 2407 | Intron | NP_004150.1 | |||
| NM_148919.3 | 2407 | Intron | NP_683720.2 | |||
| PSMB8-AS1 - PSMB8 antisense RNA 1 (head to head) | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| PSMB9 - proteasome subunit beta 9 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| TAP1 - transporter 1, ATP binding cassette subfamily B member | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000593.5 | 2407 | Missense Mutation | CCG,CGG | P,R 751 | NP_000584.2 | |
| NM_001292022.1 | 2407 | Missense Mutation | CCG,CGG | P,R 490 | NP_001278951.1 | |
| TAP2 - transporter 2, ATP binding cassette subfamily B member | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||