Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTGTATCCTTATTGTTCTTTAT[A/G]TGCTATTGGAACACAGTAATAATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608618 MIM: 606267 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM3C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM3C - family with sequence similarity 3 member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040020.1 | 937 | Intron | NP_001035109.1 | |||
NM_014888.2 | 937 | Intron | NP_055703.1 | |||
XM_011515736.2 | 937 | Intron | XP_011514038.1 | |||
XM_011515737.2 | 937 | Silent Mutation | CAC,CAT | H,H 199 | XP_011514039.1 |
WNT16 - Wnt family member 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |