Search
Search

NRL
PCK2GGATCTCACCTTTCTCCAGAGTTCT[C/G]CCCTGGTGAATGCAAACTTGGGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
NRL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| NRL - neural retina leucine zipper | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_006177.3 | Intron | NP_006168.1 | ||||
| XM_005267708.4 | Intron | XP_005267765.1 | ||||
| XM_005267709.3 | Intron | XP_005267766.1 | ||||
| XM_005267710.3 | Intron | XP_005267767.1 | ||||
| XM_011536801.2 | Intron | XP_011535103.2 | ||||
| XM_011536802.1 | Intron | XP_011535104.1 | ||||
| XM_011536804.2 | Intron | XP_011535106.1 | ||||
| XM_011536805.2 | Intron | XP_011535107.1 | ||||
| XM_011536806.2 | Intron | XP_011535108.2 | ||||
| XM_017021350.1 | Intron | XP_016876839.1 | ||||
| PCK2 - phosphoenolpyruvate carboxykinase 2, mitochondrial | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001018073.2 | Intron | NP_001018083.2 | ||||
| NM_001291556.1 | Intron | NP_001278485.1 | ||||
| NM_001308054.1 | Intron | NP_001294983.1 | ||||
| NM_004563.3 | Intron | NP_004554.3 | ||||
| XM_006720158.2 | Intron | XP_006720221.1 | ||||