Search
Search

ARHGAP4
NAA10
RENBPCAGAAGCTGCCCACCTGGGGCCAGG[A/G]AAGGCCATGGTAGAAGTAGTATTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
ARHGAP4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| ARHGAP4 - Rho GTPase activating protein 4 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| NAA10 - N(alpha)-acetyltransferase 10, NatA catalytic subunit | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001256119.1 | 359 | Missense Mutation | CCC,TCC | P,S 37 | NP_001243048.1 | |
| NM_001256120.1 | 359 | Missense Mutation | CCC,TCC | P,S 37 | NP_001243049.1 | |
| NM_003491.3 | 359 | Missense Mutation | CCC,TCC | P,S 37 | NP_003482.1 | |
| RENBP - renin binding protein | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_002910.5 | 359 | Intron | NP_002901.2 | |||
| XM_017029698.1 | 359 | Intron | XP_016885187.1 | |||