Search
Search

STAG2AGCTCACCCTCCTTTTATATAAACT[A/T]CAGTCATGTGCTGAATAATGACGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
STAG2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| STAG2 - stromal antigen 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001042749.2 | Intron | NP_001036214.1 | ||||
| NM_001042750.1 | Intron | NP_001036215.1 | ||||
| NM_001042751.1 | Intron | NP_001036216.1 | ||||
| NM_001282418.1 | Intron | NP_001269347.1 | ||||
| NM_006603.4 | Intron | NP_006594.3 | ||||
| XM_005262357.2 | Intron | XP_005262414.1 | ||||
| XM_005262358.2 | Intron | XP_005262415.1 | ||||
| XM_005262359.3 | Intron | XP_005262416.1 | ||||
| XM_005262360.2 | Intron | XP_005262417.1 | ||||
| XM_005262361.2 | Intron | XP_005262418.1 | ||||
| XM_006724727.1 | Intron | XP_006724790.1 | ||||
| XM_006724728.3 | Intron | XP_006724791.1 | ||||
| XM_011531253.2 | Intron | XP_011529555.1 | ||||
| XM_017029232.1 | Intron | XP_016884721.1 | ||||
| XM_017029233.1 | Intron | XP_016884722.1 | ||||
| XM_017029234.1 | Intron | XP_016884723.1 | ||||
| XM_017029235.1 | Intron | XP_016884724.1 | ||||