Search
Search

ALPLTGGAAAACAGGTGGCCAGGGTTTAT[T/C]GCAGCCTTGGTATGAAGTTGAGTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
ALPL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| ALPL - alkaline phosphatase, liver/bone/kidney | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000478.5 | Intron | NP_000469.3 | ||||
| NM_001127501.3 | Intron | NP_001120973.2 | ||||
| NM_001177520.2 | Intron | NP_001170991.1 | ||||
| XM_005245818.1 | Intron | XP_005245875.1 | ||||
| XM_006710546.2 | Intron | XP_006710609.1 | ||||
| XM_017000903.1 | Intron | XP_016856392.1 | ||||