Search
Search

MASP2
TARDBPGTGGGCAAAGATGACTGTCACTCTC[G/A]TGGTTTATGTCCCCTTGAGTCAATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
MASP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| MASP2 - mannan binding lectin serine peptidase 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_006610.3 | 2303 | UTR 3 | NP_006601.2 | |||
| NM_139208.2 | 2303 | Intron | NP_631947.1 | |||
| XM_017000097.1 | 2303 | Intron | XP_016855586.1 | |||
| TARDBP - TAR DNA binding protein | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_007375.3 | 2303 | Intron | NP_031401.1 | |||
| XM_017000863.1 | 2303 | Intron | XP_016856352.1 | |||
| XM_017000864.1 | 2303 | Intron | XP_016856353.1 | |||
| XM_017000865.1 | 2303 | Intron | XP_016856354.1 | |||
| XM_017000866.1 | 2303 | Intron | XP_016856355.1 | |||
| XM_017000867.1 | 2303 | Intron | XP_016856356.1 | |||
| XM_017000868.1 | 2303 | Intron | XP_016856357.1 | |||