Search
Search

OTUD7BAAAGGATGGGGGTAGGTTTCCAGCA[T/C]GGACTTGACGTAGCTGTTCAAAATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
OTUD7B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| OTUD7B - OTU deubiquitinase 7B | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_020205.3 | 348 | Missense Mutation | CAT,CGT | H,R 48 | NP_064590.2 | |
| XM_011509782.1 | 348 | Missense Mutation | CAT,CGT | H,R 48 | XP_011508084.1 | |
| XM_011509783.2 | 348 | Missense Mutation | CAT,CGT | H,R 48 | XP_011508085.1 | |
| XM_011509784.2 | 348 | Missense Mutation | CAT,CGT | H,R 48 | XP_011508086.1 | |
| XM_011509785.2 | 348 | Missense Mutation | CAT,CGT | H,R 48 | XP_011508087.1 | |
| XM_011509786.1 | 348 | Missense Mutation | ATG,GTG | M,V 14 | XP_011508088.1 | |
| XM_011509787.1 | 348 | Missense Mutation | ATG,GTG | M,V 14 | XP_011508089.1 | |
| XM_011509788.1 | 348 | Missense Mutation | ATG,GTG | M,V 14 | XP_011508090.1 | |
| XM_017001850.1 | 348 | Missense Mutation | ATG,GTG | M,V 14 | XP_016857339.1 | |
| XM_017001851.1 | 348 | UTR 5 | XP_016857340.1 | |||