Search
Search

POLQATGGTAGTGAGTCCGTCTCACGAGA[T/G]CTGATGGTTTTATAAATGGGAGTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
POLQ PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| POLQ - DNA polymerase theta | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_199420.3 | Intron | NP_955452.3 | ||||
| XM_011512343.2 | Intron | XP_011510645.1 | ||||
| XM_011512347.2 | Intron | XP_011510649.1 | ||||
| XM_011512348.2 | Intron | XP_011510650.1 | ||||
| XM_011512352.2 | Intron | XP_011510654.1 | ||||
| XM_011512354.2 | Intron | XP_011510656.1 | ||||
| XM_017005565.1 | Intron | XP_016861054.1 | ||||