Search
Search

GRM7
GRM7-AS1GAGTGATCCACCTGCTTCAGGTTCT[C/G]AAGTAGCTGGGATTGCAGGTATCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
GRM7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| GRM7 - glutamate metabotropic receptor 7 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_000844.3 | Intron | NP_000835.1 | ||||
| NM_181874.2 | Intron | NP_870989.1 | ||||
| XM_017006272.1 | Intron | XP_016861761.1 | ||||
| XM_017006273.1 | Intron | XP_016861762.1 | ||||
| GRM7-AS1 - GRM7 antisense RNA 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||