Search
Search

ARHGAP24AGTTCAAGTGAATTCAAGGCAAATT[A/-]AACTCAGTACTCCCACCAATATATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
ARHGAP24 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| ARHGAP24 - Rho GTPase activating protein 24 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001025616.2 | Intron | NP_001020787.2 | ||||
| NM_001042669.1 | Intron | NP_001036134.1 | ||||
| NM_001287805.1 | Intron | NP_001274734.1 | ||||
| NM_031305.2 | Intron | NP_112595.2 | ||||
| XM_011532300.2 | Intron | XP_011530602.1 | ||||
| XM_017008679.1 | Intron | XP_016864168.1 | ||||