Search
Search

EPHB4
ZANGCCCTCCATCTCTGCATCCTTCTGC[G/A]ACCCAAATATGCTGTACCCAACTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||
Literature Links: |
EPHB4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap |
|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
| AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
| EUR - Not Available | ||
| AMR - Not Available |
| EPHB4 - EPH receptor B4 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_004444.4 | Intron | NP_004435.3 | ||||
| XM_017011816.1 | Intron | XP_016867305.1 | ||||
| ZAN - zonadhesin (gene/pseudogene) | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||