Search
Search

LOC100421494
SOWAHBCCTTAACCTCTGCAAGTTTTAGTGA[C/T]AAACCTGCATCAACCCTTGGGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100421494 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| LOC100421494 - microtubule affinity regulating kinase 2 pseudogene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| SOWAHB - sosondowah ankyrin repeat domain family member B | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001029870.1 | 1641 | Silent Mutation | TTA,TTG | L,L 547 | NP_001025041.1 | |