Search
Search

KANSL1
KANSL1-AS1AACATCTGCACCTAGGTTTCTGCGA[A/G]TTTCCATTTGTACTCCTAAAATCAC
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||
Literature Links: |
KANSL1 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
| Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
| EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
| SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
| AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
| EUR - Not Available | |||||
| AMR - Not Available |
| KANSL1 - KAT8 regulatory NSL complex subunit 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001193465.1 | Intron | NP_001180394.1 | ||||
| NM_001193466.1 | Intron | NP_001180395.1 | ||||
| NM_015443.3 | Intron | NP_056258.1 | ||||
| XM_006721823.1 | Intron | XP_006721886.1 | ||||
| XM_006721824.3 | Intron | XP_006721887.1 | ||||
| XM_011524628.2 | Intron | XP_011522930.1 | ||||
| XM_011524629.2 | Intron | XP_011522931.1 | ||||
| XM_011524630.2 | Intron | XP_011522932.1 | ||||
| XM_011524631.2 | Intron | XP_011522933.1 | ||||
| XM_011524632.2 | Intron | XP_011522934.1 | ||||
| XM_017024488.1 | Intron | XP_016879977.1 | ||||
| XM_017024489.1 | Intron | XP_016879978.1 | ||||
| KANSL1-AS1 - KANSL1 antisense RNA 1 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||