Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCCTGCCTCTACTAGGCGCCTT[A/G]GCTCACTATGGGGAACCACTTGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600732 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL4D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARL4D - ADP ribosylation factor like GTPase 4D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001661.3 | 102 | UTR 5 | NP_001652.2 | |||
XM_011524782.2 | 102 | UTR 5 | XP_011523084.1 |