Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAGAAGCCTCAGTGGTGCGTGAA[G/T]ATTTCTAAACAGAGGCAGAGAATAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616383 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UGT3A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UGT3A1 - UDP glycosyltransferase family 3 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171873.1 | 1649 | Intron | NP_001165344.1 | |||
NM_152404.3 | 1649 | UTR 3 | NP_689617.3 | |||
XM_005248243.4 | 1649 | UTR 3 | XP_005248300.1 | |||
XM_011513957.2 | 1649 | UTR 3 | XP_011512259.1 | |||
XM_011513958.2 | 1649 | UTR 3 | XP_011512260.1 | |||
XM_011513959.2 | 1649 | UTR 3 | XP_011512261.1 | |||
XM_011513961.2 | 1649 | UTR 3 | XP_011512263.1 |