Search
Search

HOXD-AS2
HOXD10
HOXD3
HOXD8
HOXD9
LOC100129455TCCTGCATTCTTCAGCCTCCCCCTG[A/C]GCCCAGGGCTGACGTCTATCAAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOXD-AS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| HOXD-AS2 - HOXD cluster antisense RNA 2 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| HOXD10 - homeobox D10 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| HOXD3 - homeobox D3 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| HOXD8 - homeobox D8 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001199746.1 | Intron | NP_001186675.1 | ||||
| NM_001199747.1 | Intron | NP_001186676.1 | ||||
| NM_019558.3 | Intron | NP_062458.1 | ||||
| HOXD9 - homeobox D9 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| LOC100129455 - uncharacterized LOC100129455 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||