Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612121 | ||||||||||||||||||||
Literature Links: |
PNPLA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PNPLA1 - patatin like phospholipase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145716.2 | Intron | NP_001139188.1 | ||||
NM_001145717.1 | Intron | NP_001139189.2 | ||||
NM_173676.2 | Intron | NP_775947.2 | ||||
XM_011514519.2 | Intron | XP_011512821.1 | ||||
XM_011514520.2 | Intron | XP_011512822.1 | ||||
XM_017010776.1 | Intron | XP_016866265.1 | ||||
XM_017010777.1 | Intron | XP_016866266.1 | ||||
XM_017010778.1 | Intron | XP_016866267.1 | ||||
XM_017010779.1 | Intron | XP_016866268.1 |