Search
Search

Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
|||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OTUD7B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
| OTUD7B - OTU deubiquitinase 7B | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_020205.3 | 2522 | Intron | NP_064590.2 | |||
| XM_011509782.1 | 2522 | Intron | XP_011508084.1 | |||
| XM_011509783.2 | 2522 | Intron | XP_011508085.1 | |||
| XM_011509784.2 | 2522 | Intron | XP_011508086.1 | |||
| XM_011509785.2 | 2522 | UTR 5 | XP_011508087.1 | |||
| XM_011509786.1 | 2522 | Intron | XP_011508088.1 | |||
| XM_011509787.1 | 2522 | Intron | XP_011508089.1 | |||
| XM_011509788.1 | 2522 | Intron | XP_011508090.1 | |||
| XM_017001850.1 | 2522 | Intron | XP_016857339.1 | |||
| XM_017001851.1 | 2522 | Intron | XP_016857340.1 | |||
Set Membership: |
|