Search
Search

MBOAT2TTCATTCCTATGGCCTTCTATCGTG[C/T]CCCTTCTGGTTCAGAGGAGCAGTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
|||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MBOAT2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
| MBOAT2 - membrane bound O-acyltransferase domain containing 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001321265.1 | 7243 | UTR 3 | NP_001308194.1 | |||
| NM_001321266.1 | 7243 | UTR 3 | NP_001308195.1 | |||
| NM_001321267.1 | 7243 | UTR 3 | NP_001308196.1 | |||
| NM_138799.3 | 7243 | UTR 3 | NP_620154.2 | |||
| XM_011510313.1 | 7243 | Intron | XP_011508615.1 | |||
| XM_011510314.2 | 7243 | Intron | XP_011508616.1 | |||
Set Membership: |
|