Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NEMP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| NEMP1 - nuclear envelope integral membrane protein 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001130963.1 | Intron | NP_001124435.1 | ||||
| NM_015257.2 | Intron | NP_056072.2 | ||||
| XM_011538056.2 | Intron | XP_011536358.1 | ||||
| XM_011538057.2 | Intron | XP_011536359.1 | ||||
| XM_011538058.2 | Intron | XP_011536360.1 | ||||
| XM_011538059.1 | Intron | XP_011536361.1 | ||||
| XM_011538060.2 | Intron | XP_011536362.1 | ||||
| XM_017019077.1 | Intron | XP_016874566.1 | ||||
| XM_017019078.1 | Intron | XP_016874567.1 | ||||
| XM_017019079.1 | Intron | XP_016874568.1 | ||||