Search
Search

NCOA1AACAACAAAAAATTTAATGCTGTAG[A/G]GAAAGGATATTGAAAATGGATATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCOA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| NCOA1 - nuclear receptor coactivator 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_003743.4 | Intron | NP_003734.3 | ||||
| NM_147223.2 | Intron | NP_671756.1 | ||||
| NM_147233.2 | Intron | NP_671766.1 | ||||
| XM_005264625.1 | Intron | XP_005264682.1 | ||||
| XM_005264626.1 | Intron | XP_005264683.1 | ||||
| XM_005264628.1 | Intron | XP_005264685.1 | ||||
| XM_017005168.1 | Intron | XP_016860657.1 | ||||
| XM_017005169.1 | Intron | XP_016860658.1 | ||||
Set Membership: |
|