Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACCCACGGGACAGACTGGGGAAG[A/C]GGCCCAGGGTCCATCAAGTAAACCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613066 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRT18P59 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KRT18P59 - keratin 18 pseudogene 59 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PKNOX2 - PBX/knotted 1 homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022062.2 | 358 | Intron | NP_071345.2 | |||
XM_005271642.2 | 358 | Intron | XP_005271699.1 | |||
XM_005271643.2 | 358 | Intron | XP_005271700.1 | |||
XM_006718894.2 | 358 | Intron | XP_006718957.1 | |||
XM_011542944.2 | 358 | Intron | XP_011541246.1 | |||
XM_011542945.2 | 358 | Intron | XP_011541247.1 | |||
XM_011542946.1 | 358 | Intron | XP_011541248.1 | |||
XM_011542947.2 | 358 | Intron | XP_011541249.1 | |||
XM_017018110.1 | 358 | UTR 5 | XP_016873599.1 | |||
XM_017018111.1 | 358 | Intron | XP_016873600.1 | |||
XM_017018112.1 | 358 | Intron | XP_016873601.1 |
TMEM218 - transmembrane protein 218 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080546.2 | 358 | Intron | NP_001074015.1 | |||
NM_001258238.1 | 358 | Intron | NP_001245167.1 | |||
NM_001258239.1 | 358 | Intron | NP_001245168.1 | |||
NM_001258240.1 | 358 | Intron | NP_001245169.1 | |||
NM_001258241.1 | 358 | Intron | NP_001245170.1 | |||
NM_001258242.1 | 358 | Intron | NP_001245171.1 | |||
NM_001258243.1 | 358 | Intron | NP_001245172.1 | |||
NM_001258244.1 | 358 | Intron | NP_001245173.1 | |||
NM_001258245.1 | 358 | Intron | NP_001245174.1 | |||
NM_001258246.1 | 358 | Intron | NP_001245175.1 | |||
NM_001258247.1 | 358 | Intron | NP_001245176.1 | |||
XM_006718779.3 | 358 | Intron | XP_006718842.1 | |||
XM_006718780.3 | 358 | Intron | XP_006718843.1 | |||
XM_006718781.3 | 358 | Intron | XP_006718844.1 | |||
XM_006718783.3 | 358 | Intron | XP_006718846.1 | |||
XM_017017322.1 | 358 | Intron | XP_016872811.1 | |||
XM_017017323.1 | 358 | Intron | XP_016872812.1 | |||
XM_017017324.1 | 358 | Intron | XP_016872813.1 |
Set Membership: |
HapMap |