Search
Search

AFF2TTTCCAAACAAATAGTTAAATTAAA[C/T]GTGAGCTTCTGAATTGTACTTGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AFF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| AFF2 - AF4/FMR2 family member 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001169122.1 | 11313 | UTR 3 | NP_001162593.1 | |||
| NM_001169123.1 | 11313 | UTR 3 | NP_001162594.1 | |||
| NM_001169124.1 | 11313 | UTR 3 | NP_001162595.1 | |||
| NM_001169125.1 | 11313 | UTR 3 | NP_001162596.1 | |||
| NM_001170628.1 | 11313 | UTR 3 | NP_001164099.1 | |||
| NM_002025.3 | 11313 | UTR 3 | NP_002016.2 | |||
Set Membership: |
|