Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTCCACGTCTCTGTTTCCACATC[A/G]TAGCGCTCCACGCTGTTCAGCTGGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606016 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KEAP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KEAP1 - kelch like ECH associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012289.3 | 1935 | Silent Mutation | TAC,TAT | Y,Y 537 | NP_036421.2 | |
NM_203500.1 | 1935 | Silent Mutation | TAC,TAT | Y,Y 537 | NP_987096.1 | |
XM_005260173.1 | 1935 | Silent Mutation | TAC,TAT | Y,Y 537 | XP_005260230.1 | |
XM_005260174.1 | 1935 | Silent Mutation | TAC,TAT | Y,Y 537 | XP_005260231.1 | |
XM_011528452.1 | 1935 | Silent Mutation | TAC,TAT | Y,Y 537 | XP_011526754.1 |
Set Membership: |
HapMap |